Learn More
CPC Scientific H-Ala-Val-Ser-Glu-His-Gln-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110342
Encompass
Peptide found to have anabolic effect on bone formation in rats that can inhibit the rapid decline in bone formation due to denervation.ONE-LETTER SEQUENCE: AVSEHQLLHDKGKSIQDLRRRFFLHHLIAEIHTAMOLECULAR FORMULA: C180H287N57O48MOLECULAR WEIGHT:4016.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [112955-31-4], RESEARCH AREA: OsteoporosisREFERENCES: L.J. Suva et al., Science, 237, 893 (1987)
Catalog No. 50-211-0342 Supplier CPC Scientific Supplier No. PTHP005B
Peptide found to have anabolic effect on bone formation in rats that can inhibit the rapid decline in bone formation due to denervation.ONE-LETTER SEQUENCE: AVSEHQLLHDKGKSIQDLRRRFFLHHLIAEIHTAMOLECULAR FORMULA: C180H287N57O48MOLECULAR WEIGHT:4016.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [112955-31-4], RESEARCH AREA: OsteoporosisREFERENCES: L.J. Suva et al., Science, 237, 893 (1987)