Learn More
Apexbio Technology LLC HyperFluor™ 555 Goat Anti-Rabbit IgG (H+L) Antibody 5X100ul
SDP
Supplier:  Apexbio Technology LLC K33090.5
Encompass_Preferred
HyperFluor 555 Goat Anti-Rabbit IgG HplusL Antibody is an affinity-purified polyclonal antibody targeting rabbit immunoglobulin G (IgG) heavy and light chains It is designed to recognize and bind to primary antibodies raised in rabbits thereby enabling indirect detection and signal amplification HyperFluor 555 Goat Anti-Rabbit IgG HplusL Antibody exerts its biological activity primarily through specific binding to rabbit IgG (H L) and is conjugated with the HyperFluor 555 fluorescent dye (excitation/emission 555/568 nm) Based on these properties HyperFluor 555 Goat Anti-Rabbit IgG HplusL Antibody holds research potential in fluorescence microscopy immunofluorescence staining flow cytometry and related fluorescence-based assays
Catalog No. 50-000-97304