Learn More
Apexbio Technology LLC HyperFluor™ 594 Goat Anti-Human IgG (H+L) Antibody 5X100ul
SDP
Supplier:  Apexbio Technology LLC K33070.5
Encompass_Preferred
HyperFluor 594 Goat Anti-Human IgG HplusL Antibody is an affinity-purified secondary antibody that specifically targets human immunoglobulin G (IgG) recognizing both heavy and light chains (H L) Conjugated with the HyperFluor 594 fluorophore it facilitates detection of IgG via fluorescence with excitation at 590 nm and emission at 617 nm Based on these characteristics HyperFluor 594 Goat Anti-Human IgG HplusL Antibody is suitable for applications including immunocytochemistry immunohistochemistry and flow cytometry
Catalog No. 50-000-97222