Learn More
Apexbio Technology LLC HyperFluor™ 594 Goat Anti-Mouse IgG (H+L) Antibody 100ul
SDP
Supplier:  Apexbio Technology LLC K33060.1
Encompass_Preferred
HyperFluor 594 Goat Anti-Mouse IgG HplusL Antibody is a fluorescently labeled secondary antibody targeting mouse immunoglobulins (IgG heavy and light chains) HyperFluor 594 Goat Anti-Mouse IgG HplusL Antibody exerts its biological utility by specific binding to mouse IgG enabling detection of primary antibodies in immunological assays Based on these properties HyperFluor 594 Goat Anti-Mouse IgG HplusL Antibody is applicable in immunocytochemistry immunohistochemistry and flow cytometry for fluorescence-based detection of mouse-derived primary antibodies
Catalog No. 50-000-97206