Learn More
Medchemexpress LLC Hypericin-d10 | 548-04-9 | 95.3% | 514.50 | C30H6D10O8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0453S10MG
Encompass_Preferred
Hypericin-d10 is the deuterium-labeled analogue of hypericin, provided for research use as a stable isotope tracer or internal standard in analytical workflows. It is intended for quantitative and pharmacokinetic studies using techniques such as NMR, GC-MS, and LC-MS, and exhibits the characteristic photosensitive biological activity of hypericin derivatives.
  • Deuterium-labeled tracer for quantitative analysis.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • Reported purity of 95.3% supports analytical applications.
  • Solid, brown to black appearance; store under recommended conditions for stability.
  • Molecular formula C30H6D10O8 and molecular weight 514.50 aid reference and calculations.
Catalog No. 50-002-95655