Learn More
Medchemexpress LLC Hypericin-d2 | 506.46 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0453S11MG
Encompass_Preferred
Hypericin-d2 is deuterium labeled Hypericin for research use only. It is intended to be used as a tracer and internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable for NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
Catalog No. 50-003-89774