Learn More
Medchemexpress LLC HY-N2575 5mg Medchemexpress, Hypocrellin A CAS:77029-83-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25755MG
Encompass_Preferred
Medchemexpress, HY-N2575 5mg Hypocrellin A CAS:77029-83-5 Hypocrellin A, a naturally occurring PKC inhibitor, has many biological and pharmacological properties, such as antitumour, antiviral, antibacterial, and antileishmanial activities. Hypocrellin A is a promising photosensitizer for anticancer photodynamic therapy (PDT)[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2906