Learn More
Medchemexpress LLC Hypocrellin B | 123940-54-5 | 528.51 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN145350MG
Encompass_Preferred
Hypocrellin B is a pigment isolated from the fungi Hypocrella bambusae and Shiraia bambusicola and acts as an apoptosis inducer. It can be utilized as a photosensitizer for the photodynamic therapy of cancer and also exhibits antimicrobial and antileishmanial activities.
  • Apoptosis inducer
  • Photosensitizer for photodynamic therapy of cancer
  • Antimicrobial activity
  • Antileishmanial activity
  • Induces cell cytotoxicity and early apoptosis in HepG2 cells after sonodynamic therapy
  • Shows antifungal activity against C. albicans
  • Shows anti-parasite activity against Leishmania donovani
  • Inhibits tumor growth in A549 tumor bearing mice
Catalog No. 50-004-14644