Learn More
Research Products International Corp Hypoxanthine 1 KG

Supplier: Research Products International Corp H620001000.0
Molecular Formula: C5H4N4ONa
MW 158.11
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.