Learn More
Medchemexpress LLC Hypoxanthine-d2 | 697807-02-6 | 98.0% | 138.12 g/mol | C5H2D2N4O | 10 MG

Supplier: Medchemexpress LLC HYN0091S610MG
Hypoxanthine-d2 is the deuterium-labeled form of hypoxanthine, a purine derivative used in analytical and metabolomics research. It functions as a potential free radical generator and can serve as an indicator of hypoxia, and is commonly employed as a tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
- Deuterium-labeled tracer for NMR, GC-MS, and LC-MS.
- Useful as an indicator of hypoxia and in oxidative stress studies.
- Purity 98.0%.
- Available in small preweighed amounts suitable for analytical workflows.
- Recommended storage: sealed, away from moisture and light; in solvent store at -80°C up to 6 months, -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.