Learn More
Selleck Chemical LLC Hypromellose S4444-100mg
SDP
Supplier:  Selleck Chemical LLC S4444100MG
Encompass_Preferred
Hypromellose (Hydroxypropyl Methyl Cellulose HPMC Celacol HPM 5000) is a commonly employed cellulose ether used in the fabrication of hydrophilic matrices Hypromellose provides the release of a drug in a controlled manner effectively increasing the duration of release of a drug to prolong its therapeutic effect
Catalog No. 50-311-2196