Learn More
Medchemexpress LLC HY-107443 10mg Medchemexpress, I-BET762 carboxylic acid CAS:1300019-38-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10744310MG
Encompass_Preferred
Medchemexpress, HY-107443 10mg I-BET762 carboxylic acid CAS:1300019-38-8 I-BET762 carboxylic acid (Molibresib carboxylic acid) is an I-BET762-based warhead ligand for conjugation reactions of PROTAC targeting on BET. I-BET762 carboxylic acid (Molibresib carboxylic acid) is a BRD4 inhibitor with a pIC50 of 5.1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8778