Learn More
Eurofins Discoverx I-BOP, 100 µg. (92-1038)
Supplier:  Eurofins Discoverx 921038
Encompass
I-BOP is a small molecule that induces platelet aggregation and vascular smooth muscle contraction. It is a thromboxane mimetic and a potent TP receptor agonist, with an EC50 of 0.34 nM for TBXA2R. Provided as 100 μg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9583