Learn More
Medchemexpress LLC I-inositol-d | 87-89-8 | 99.5% | 181.16 g/mol | C6H11DO6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1411S110MG
Encompass_Preferred
i-Inositol-d (myo-inositol-d1) is a monodeuterated form of myo-inositol used as a stable isotope standard and research reagent for metabolic and biochemical studies. Supplied as a solid powder for analytical and mass spectrometry applications, it has a molecular weight of 181.16 g/mol and is provided at high purity for accurate quantitation.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative analysis.
  • High purity (≈99.5%) for reliable analytical results.
  • Molecular weight 181.16 g/mol; chemical formula C6H11DO6.
  • Supplied as a solid powder for convenient storage and handling.
  • Suitable for metabolic pathway studies and biochemical assays.
Catalog No. 50-002-98925