Learn More
Cayman Chemical I-OMe AG-538 25mg
SDP
Supplier:  Cayman Chemical 3509025 MG
Encompass_Preferred
An IGF-1R and PI5P4Ka inhibitor (IC50s = 3.4 and 1 µM, respectively); selectively cytotoxic to nutrient-deprived PANC-1 cells over nutrient-sufficient PANC-1 cells
Catalog No. 50-291-1532