Learn More
Medchemexpress LLC I2959 photoinitiator | 106797-53-9 | 98.67% | 224.26 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013508100G
Encompass_Preferred
I2959 is a photoinitiator widely used for the photochemical cross-linking of hydrogels. Its combination with CMA hydrogel has been utilized as a bio-ink in biological 3D printing applications. This product exhibits high free radical generation efficiency and is cytotoxic to rapidly dividing cell lines.
  • Used as a photoinitiator in photochemical cross-linking of hydrogels.
  • Applied as a bio-ink for biological 3D printing when combined with CMA hydrogel.
  • Generates free radicals efficiently.
  • Cytotoxic to rapidly dividing cell lines.
  • Suitable for in vitro encapsulation of Saos-2 cells in photochemically cross-linked CMA hydrogels.
Catalog No. 50-004-54383