Learn More
Medchemexpress LLC HY-136205 10mg , IA-Alkyne CAS:930800-38-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13620510MG
Encompass_Preferred
HY-136205 10mg IA-Alkyne CAS: 930800-38-7 IA-Alkyne (Iodoacetamide-alkyne; N-Hex-5-ynyl-2-iodo-acetamide) is a TRP channel (TRPC) agonist and has the potential for the study of respiratory infection. IA-Alkyne can be used to develop an isotopically tagged probe for quantitative cysteine-reactivity profiling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1404