Learn More
Selleck Chemical LLC IACS-010759 S8731-1G
SDP
Supplier:  Selleck Chemical LLC S87311G
Encompass_Preferred
IACS-010759 (IACS-10759) is a potent and selective oxidative phosphorylation inhibitor (IC50 10 nM) that blocks cellular respiration through inhibition of complex I
Catalog No. 50-000-24213