Learn More
Selleck Chemical LLC IACS-010759 (IACS-10759) 5mg 1570496-34-2
SDP
Supplier:  Selleck Chemical LLC S87315MG
Encompass_Preferred
IACS-010759 (IACS-10759) is a potent and selective oxidative phosphorylation inhibitor (IC50 < 10 nM) that blocks cellular respiration through inhibition of complex I. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8054