Learn More
Medchemexpress LLC IACS-8803 disodium | 2243079-36-7 | 98.8% | 50 MG
Supplier: Medchemexpress LLC HY130115A50MG
IACS-8803 disodium is a highly potent cyclic dinucleotide STING agonist with robust systemic antitumor efficacy. It is intended for research use only.
- Highly potent cyclic dinucleotide STING agonist
- Robust systemic antitumor efficacy
- Shows superior antitumor activity in mice bearing bilateral B16-OVA melanomas
- Targets STING pathway
- Involves immunology/inflammation pathway