Learn More
Medchemexpress LLC IACS-8803 disodium | 2243079-36-7 | 99.97% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY130115A10MM/1ML
Encompass_Preferred
IACS-8803 disodium is a highly potent cyclic dinucleotide STING agonist that exhibits robust systemic antitumor efficacy.
  • Highly potent cyclic dinucleotide STING agonist
  • Exhibits robust systemic antitumor efficacy
Catalog No. 50-003-38379