Learn More
Medchemexpress LLC IACS-8803 disodium | 2243079-36-7 | C20H21FN10Na2O9P2S2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY130115A10MG
Encompass_Preferred
IACS-8803 disodium is a cyclic dinucleotide stimulator of interferon genes (STING) agonist described by investigators at the Institute for Applied Cancer Science (MD Anderson). Supplied as the disodium salt for research use, it shows robust systemic antitumor efficacy in preclinical studies and includes documented solubility and formulation guidance for in vitro and in vivo experiments.
  • Potent activation of the STING pathway demonstrated in preclinical studies.
  • Water-soluble disodium salt with documented solubility in DMSO, water, and PBS.
  • Suggested in vivo formulations and dosing vehicles provided for clear solution preparation.
  • High chemical purity suitable for laboratory research applications.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-58780