Learn More
Medchemexpress LLC HY-112164 1mg Medchemexpress, IACS-8968 CAS:2144425-14-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1121641MG
Encompass_Preferred
Medchemexpress, HY-112164 1mg IACS-8968 CAS:2144425-14-7 IACS-8968 (IDO/TDO Inhibitor) is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9044