Learn More
Medchemexpress LLC HY-112164A 1mg Medchemexpress, IACS-8968 (R-enantiomer) CAS:2239305-67-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112164A1MG
Encompass_Preferred
Medchemexpress, HY-112164A 1mg IACS-8968 (R-enantiomer) CAS:2239305-67-8 IACS-8968 (R-enantiomer) is the R-enantiomer of IACS-8968. IACS-8968 is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9045