Learn More
Medchemexpress LLC IACS-9571 hydrochloride | 2319611-93-1 | 98.3% | 679.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC 1MGHY102000B
Encompass_Preferred
IACS-9571 hydrochloride (ASIS-P040) is a potent and selective inhibitor of TRIM24 and BRPF1. It shows excellent cellular potency with EC50 of 50 nM and potent activities against a panel of 32 bromodomains. It is also highly selective against BRPF2 and BRPF3, and does not interact with the BET sub-family of bromodomains.
  • Potent and selective TRIM24 and BRPF1 inhibitor
  • Excellent cellular potency (EC50 of 50 nM)
  • Potent activities against 32 bromodomains
  • High selectivity against BRPF2 and BRPF3
  • No interaction with BET sub-family bromodomains
Catalog No. 50-003-65453