Learn More
Medchemexpress LLC IACS-9571 hydrochloride | 2319611-93-1 | 98.3% | 679.22 g/mol | C32H43ClN4O8S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY102000B10MG
Encompass_Preferred
IACS-9571 hydrochloride is a selective, low-nanomolar inhibitor of the TRIM24 and BRPF1 bromodomains used as a research tool to probe bromodomain biology and epigenetic regulation. It is supplied as the hydrochloride salt and is characterized by low-nanomolar binding (IC50 ≈ 8 nM for TRIM24; Kd values in the low nanomolar range for TRIM24 and BRPF1).
  • Selective dual TRIM24/BRPF1 bromodomain inhibition
  • Low-nanomolar potency (IC50 ≈ 8 nM)
  • High purity suitable for research (~98.3%)
  • Available in small research pack sizes for in vitro work
  • Offered as solid and as 10 mM solutions in DMSO
Catalog No. 50-002-47536