Learn More
Genetex Inc Iba1 Monoclonal Antibody [MSVA-955M] HistoMAX™ (GTX04465) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04465
Encompass_Preferred
The Iba1 antibody [MSVA-955M] HistoMAX™ (GTX04465) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human epithelioid malignant mesothelioma tissue using GTX04465 Iba1 antibody [MSVA-955M] HistoMAX. Malignant mesothelioma epitheloid subtype containing numerous Iba1 positive inflammatory cells.
Catalog No. NC3667750