Learn More
Selleck Chemical LLC Ibandronate sodium monohydrate S3148-50mg
SDP
Supplier:  Selleck Chemical LLC S314850MG
Encompass_Preferred
Ibandronate (BM-21 0955 BM-210955 RPR-102289A) is a highly potent nitrogen-containing bisphosphonate used for the treatment of osteoporosis
Catalog No. 50-310-8239