Learn More
Selleck Chemical LLC Iberdomide S8760-1G
SDP
Supplier:  Selleck Chemical LLC S87601G
Encompass_Preferred
Iberdomide (CC-220) is a novel oral immunomodulatory compound that targets cereblon part of the CRL4CRBN E3 ubiquitin ligase complex with an IC50 of 60 nM in a competitive TR-FRET assay Iberdomide (CC-220) induces apoptosis with antitumor and immunostimulatory activities
Catalog No. 50-000-24094