Learn More
Selleck Chemical LLC Iberiotoxin TFA-E0015-5MG
SDP
Supplier:  Selleck Chemical LLC E00155MG
Encompass_Preferred
Iberiotoxin TFA(IbTX) is a toxin purified from the scorpion Buthus tamulus Iberiotoxin is a potent blocker of the large conductance Ca(2 )-activated K channel with a Kd of 1 nM
Catalog No. 50-313-4641