Learn More
Cayman Chemical IBET762 Inhibitors
SDP
Supplier:  Cayman Chemical 1067610 MG
Encompass_Preferred
A synthetic compound which interacts with BET proteins with high-affinity (Kd = 32.5-42.5 nM); disrupts binding of BET proteins with acetylated histones, suppressing the expression of specific inflammatory genes involved in sepsis and endotoxic shock
Catalog No. 50-288-8302