Learn More
Medchemexpress LLC Ibezapolstat | 1275582-97-2 | 98.0% | 10 MG

Supplier: Medchemexpress LLC HY12835710MG
Ibezapolstat (ACX-362E) is a first-in-class, orally active DNA polymerase IIIC (pol IIIC) inhibitor developed for the research of C. difficile infection (CDI). It is for research use only.
- Inhibits DNA pol IIIC from aerobic and low G+C Gram-positive bacteria
- Active against a broad spectrum of C. difficile pathogens
- MIC range of 1-8 μg/mL for 104 C. difficile clinical isolates
- Poorly absorbed and nontoxic in hamster C. difficile-associated disease model
- Strong anti-C. difficile properties in hamster CDAD model, protecting infected animals for up to 5 days
- Appears as a white to off-white solid
- Soluble in DMSO (62.5 mg/mL)
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.