Learn More
Medchemexpress LLC Ibezapolstat | 1275582-97-2 | 98.0% | 423.30 g/mol | C18H20Cl2N6O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1283575MG
Encompass_Preferred
Ibezapolstat is a research-grade small molecule inhibitor of bacterial DNA polymerase IIIC, developed as an orally active antibiotic candidate for studies of Clostridioides difficile and bacterial DNA/RNA synthesis. Supplied as a powder for in vitro and in vivo research, it is provided in low-milligram quantities for experimental dosing and formulation work.
  • First-in-class oral DNA polymerase IIIC inhibitor.
  • Suitable for in vitro and in vivo antibiotic research.
  • Supplied as a powder for ease of dissolution and formulation.
  • Provided in small mg packaging for dose-ranging studies.
  • Characterized with CAS 1275582-97-2 and defined molecular formula.
Catalog No. 50-002-64582