Learn More
Medchemexpress LLC Ibezapolstat (ACX-362E) | 1275582-97-2 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128357100MG
Encompass_Preferred
Ibezapolstat (ACX-362E) is a first-in-class, orally active DNA polymerase IIIC inhibitor developed for the research of *C. difficile* infection (CDI). It demonstrates antibacterial activity against a broad spectrum of *C. difficile* pathogens by inhibiting DNA pol IIIC from aerobic and low G+C Gram-positive bacteria. This compound is poorly absorbed, suggesting potential for high local concentrations at the site of infection in the colon, and has shown strong anti-*C. difficile* properties in vivo.
  • First-in-class, orally active DNA polymerase IIIC inhibitor
  • Developed for *C. difficile* infection (CDI) research
  • Exhibits antibacterial activity against *C. difficile* pathogens
  • Inhibits DNA pol IIIC from aerobic and low G+C Gram-positive bacteria
  • Poorly absorbed in vivo, promoting local concentration at infection site
  • Demonstrates strong anti-*C. difficile* properties in animal models
Catalog No. 50-003-40819