Learn More
Medchemexpress LLC HY-16743 10mg , Ibiglustat CAS:1401090-53-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1674310MG
Encompass_Preferred
HY-16743 10mg Ibiglustat CAS: 1401090-53-6 Ibiglustat (Venglustat) is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. Ibiglustat can be used for the research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9769