Learn More
Medchemexpress LLC HY-12318 50mg , IBMX IMX;Isobutylmethylxanthine;Methylisobutylxanthine;NSC165960;SC2964 CAS:28822-58-4 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY1231850MG
Encompass_Preferred
Medchemexpress, HY-12318 50mg IBMX IMX;Isobutylmethylxanthine;Methylisobutylxanthine;NSC165960;SC2964 CAS:28822-58-4 Formula:C10H14N4O2 IC50: 6.5±1.2 μM(PDE3), 26.3±3.9 μM (PDE4), 31.7±5.3 μM (PDE5) Purity:98% IBMX is a broad-spectrum phosphodiesterase ( PDE ) inhibitor, with IC 50 of 6.5±1.2, 26.3±3.9 and 31.7±5.3 μM for PDE3 , PDE4 and PDE5 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3700