Learn More
Medchemexpress LLC Ibotenic acid | 2552-55-8 | 50 MG

Supplier: Medchemexpress LLC HYN231150MG
Ibotenic acid, also known as (RS)-Ibotenic acid or DL-Ibotenic acid, is an iGluR Agonist. It displays agonist activity at both the N-methyl-D-aspartate (NMDA) and trans-ACPD or metabolotropic quisqualate (Qm) receptor sites. Its agonist activity at NMDA receptors, which is linked to its neurotoxicity, makes it valuable for neurobiological research, particularly in studies concerning neurological conditions. This product is intended for research purposes only and is not for sale to patients.
- Agonist activity at NMDA and trans-ACPD/metabotropic quisqualate (Qm) receptor sites
- Purity: 99.34%
- Molecular weight: 158.11
- Formula: C₅H₆N₂O₄
- Appearance: Solid, light yellow to yellow color
- Source: Microorganisms, Amanita mushrooms
- Toxic to cortical neurons with an EC50 = 77.3 ± 8 μM
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.