Learn More
Medchemexpress LLC Ibotenic acid | 2552-55-8 | MFCD00069294 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN2311
Ibotenic acid is an iGluR Agonist that exhibits activity at both N-methyl-D-aspartate (NMDA) and trans-ACPD or metabolotropic quisqualate (Qm) receptor sites. It is toxic to cortical neurons, with an EC50 of 77.3±8 μM as measured by the release of lactate dehydrogenase to the culture media after 5 minutes of exposure. This compound is suitable for research use.
- Functions as an iGluR agonist
- Activates N-methyl-D-aspartate (NMDA) and trans-ACPD/Qm receptor sites
- Molecular weight: 158.11
- Chemical formula: C5H6N2O4
- Appears as a solid, light yellow to yellow in color
- Sourced from Amanita mushrooms
- Purity of 99.34%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.