Learn More
Medchemexpress LLC Ibotenic acid | 2552-55-8 | MFCD00069294 | 99.3% | 158.11 g/mol | C5H6N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN231110MG
Encompass_Preferred
Ibotenic acid is a research-grade excitatory amino acid agonist that activates ionotropic and metabotropic glutamate receptors, including NMDA and trans-ACPD/quisqualate (Qm) sites. Supplied as a solid for laboratory research, it is used in receptor pharmacology, neurotoxicity models, and in vitro assays.
  • Agonist at NMDA and Qm receptor sites.
  • High purity for reliable experimental results.
  • Molecular formula C5H6N2O4; molecular weight 158.11 g/mol.
  • Supplied in small milligram quantities suitable for research (for example, 10 mg).
  • White solid; handle with appropriate safety precautions due to neuroactive properties.
Catalog No. 50-002-89331