Learn More
Abcam Ibotenic acid, e x citoto x ic agonist, 5MG
Supplier:  Abcam AB1200415MG
Encompass_Preferred
MW 158.11 Da, Purity >98%. Neuroexcitatory amino acid originally isolated from Amanita species. Neurotoxin often used to model cognitive dysfunctions. NMDA and metabotropic receptor agonist.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7905