Learn More
Medchemexpress LLC Ibrolipim (NO-1886) | 133208-93-2 | 99.5% | 451.25 g/mol | C19H20BrN2O4P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11754910MM 1ML
Encompass_Preferred
Ibrolipim (NO-1886) is an orally active lipoprotein lipase (LPL)-promoting small molecule used in research on lipid metabolism and cardiovascular disease. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as solid quantities for custom preparation. Reported molecular weight is 451.25 g/mol and CAS is 133208-93-2.
  • Ready-to-use 10 mM solution in DMSO, 1 mL
  • High reported purity (~99.5%)
  • Suitable for in vitro and in vivo lipid metabolism studies
  • Also available as solids for custom solution preparation
  • Characterized compound with CAS 133208-93-2 and known molecular weight
Catalog No. 50-002-26674