Learn More
Medchemexpress LLC Ibrutinib-biotin | 1599432-18-4 | MFCD30496724 | 99.7% | 1097.37 | C56H80N12O9S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10034210MG
Encompass_Preferred
Ibrutinib-biotin is a biotinylated chemical probe derived from the BTK inhibitor ibrutinib. The probe links ibrutinib to a biotin tag via a long-chain linker, preserving low-nanomolar inhibitory potency against Bruton's tyrosine kinase while enabling affinity capture and target engagement studies. It is supplied as a high-purity solid and is also available as a DMSO solution for biochemical assays.
  • Biotin tag enables affinity capture and pull-down experiments.
  • Retains low-nanomolar BTK inhibitory potency for target engagement.
  • High purity suitable for biochemical and proteomic workflows.
  • Available as solid and as DMSO solution for assay flexibility.
  • Useful for mechanism-of-action studies and probe-based profiling.
Catalog No. 50-002-15063