Learn More
Bioworld IBS/Glycerol/Fish Gelatin, 1 L
Supplier:  Bioworld 207200051
Encompass_Preferred
IBS/Glycerol/Fish Gelatin IBS Buffered fish gelatin 5% blocks, diluents and coating solutions are supplied as 10X concentrates. Technical Information:pH range: 7.0 (+/- 0.1)
Catalog No. 50-198-405