Learn More
Ibt Bioservices IBT Ebola virus-like particles (Ebola Virus VLP), Cat # 0550-001, Ebola Virus, Ebola virus-like particles, Zaire, Column chromatography, Western Blot, Liquid, Storage: -80°C
SDP
Supplier:  Ibt Bioservices 0550001
Encompass_Preferred
Virus-like Particles (VLP’s) expressing recombinant Ebola virus (EBOV) glycoprotein (GP), nucleoprotein (NP), and matrix protein (VP40). These VLP’s are produced in Sf9 insect cells through infection with a recombinant baculovirus. Since these VLP’s mimic Ebola virus but do not contain genetic material, thus are not infectious and make an ideal candidate as a vaccine and also as a tool to enhance filovirus research.Supplied in PBS (supplemented with arginine and glutamic acid). Store at 2-3 weeks at -20°C, -80°C long term
Catalog No. NC3958472