Learn More
Ibt Bioservices IBT Mouse anti-Marburg Virus VP40 mAb (1D1), Cat # 0203-012, Mouse, Monoclonal, Marburg Virus, Marburg VP40, Angola, Protein A/G, ELISA, Western Blot, Liquid, Storage: -80°C
SDP
Supplier:  Ibt Bioservices 0203012
Encompass_Preferred
Protein G purified mouse monoclonal antibody reactive to MMARV VP40. The antibody detects VP40 in virus-like particles (VLP) and recombinant protein.Supplied in PBS in frozen format with no preservatives added. Store at +4°C for 2-3 weeks; -80°C for long-term
Catalog No. NC3956606