Learn More
Selleck Chemical LLC Ibudilast 25mg 50847-11-5 KC-404, AV411, MN166
SDP
Supplier:  Selleck Chemical LLC S483725MG
Encompass_Preferred
Ibudilast (KC-404, AV411, MN166) is a relatively non-selective phosphodiesterase inhibitor with anti-inflammatory and neuroprotective activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8206