Learn More
Medchemexpress LLC Ibuprofen-13C,d3 | 1261394-40-4 | C1213CH15D3O2 | 1 MG
Supplier: Medchemexpress LLC HY78131S11MG
Ibuprofen-13C,d3 is the 13C- and deuterium labeled Ibuprofen. Ibuprofen is an anti-inflammatory agent targeting COX-1 and COX-2 with IC50s of 13 μM and 370 μM, respectively. It serves as a valuable tool in research.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.