Learn More
Medchemexpress LLC Ibuprofen L-lysine | 57469-77-9 | MFCD01716155 | 99.9% | 352.5 g/mol | C19H32N2O4 | 100 MG

Supplier: Medchemexpress LLC HY100586100MG
Ibuprofen L-lysine is the L-lysine salt of ibuprofen provided for research use. It functions as a nonsteroidal anti-inflammatory agent and a nitric oxide donor, with reported selective COX-1 inhibitory activity (IC50 ≈13 μM). The compound is used as an analytical standard and in preclinical studies of pain, inflammation, and related cellular responses.
- High purity analytical standard (99.9%).
- Molecular weight 352.5 g/mol.
- Chemical formula C19H32N2O4.
- Intended for research and analytical applications.
- Available in small pack sizes, including 100 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.