Learn More
Medchemexpress LLC Ibuprofen L-lysine | 57469-77-9 | MFCD01716155 | 99.9% | 352.5 g/mol | C19H32N2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10058650MG
Encompass_Preferred
Ibuprofen L-lysine is the lysine salt of ibuprofen supplied for research and analytical applications. It functions as a nonsteroidal anti-inflammatory agent and a nitric oxide donor and is commonly used in studies of pain, inflammation, and pharmacology. The compound is offered in small, measured pack sizes and is characterized for laboratory use.
  • High purity specification (99.91%).
  • CAS number 57469-77-9 for precise identification.
  • Lysine salt form improves water solubility compared with the free acid.
  • Available in multiple small pack sizes suitable for analytical assays.
  • Intended for research use and not for human therapeutic use.
Catalog No. 50-002-13644