Learn More
Waters Corp Ibuprofen Standard, 3 g
Supplier:  Waters Corp 186008061
Encompass_Preferred
This Ibuprofen Standard is used to assess the reproducibility of Waters ACQUITY UPC2 Systems or SFC systems and to verify system performance.
  • Designed for system reproducibility assessment.
  • Verifies system performance.
  • Contains 3 g of ibuprofen.
Catalog No. 50-239-1375